missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR87 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118816
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GPR87/GPR95.
G protein-coupled receptors play a role in cell communication. They are characterized by an extracellular N terminus, 7 transmembrane regions, and an intracellular C terminus. GPR87 has been reported to be expressed in human prostate and rat stomach and fetal tissues. ESTs have been isolated from B-cell/lung/testis, breast, heart, jaw, prostate, and testis libraries.
Specifications
| GPR87 | |
| Polyclonal | |
| Unconjugated | |
| GPR87 | |
| FKSG78; G protein-coupled receptor 87; G protein-coupled receptor 95; GPR87; GPR95; G-protein coupled receptor 87; G-protein coupled receptor 95; KPG_002; MGC131898; orphan GPCR 87 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 53836, 84111 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q99MT7, Q9BY21 | |
| GPR87 | |
| A synthesized peptide derived from human GPR87(Accession Q9BY21), corresponding to amino acid residues T299-V349. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction