missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR88 Monoclonal Antibody (1039830)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA560911
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
GPR88 is an Orphan-A GPCR with an unknown ligand. GPR88 expression has been documented almost exclusively in the striatum of the brain and not in any peripheral tissues. ESTs have been isolated from diseased brain, ear, and kidney libraries.
Specifications
| GPR88 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9GZN0 | |
| GPR88 | |
| Human GPR88 synthetic peptide. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| 1039830 | |
| Unconjugated | |
| GPR88 | |
| AW061286; COCPMR; G protein coupled receptor 88; G protein-coupled receptor 88; Gpr88; G-protein coupled receptor 88; Probable G-protein coupled receptor 88; Strg; Striatum-specific G-protein coupled receptor | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 54112 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction