missing translation for 'onlineSavingsMsg'
Learn More

GPRC5A/RAI3 Antibody (GPRC5A/7893), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18512039 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18512039 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18512039 Supplier Novus Biologicals Supplier No. NBP324315AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GPRC5A/RAI3 Monoclonal antibody specifically detects GPRC5A/RAI3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPRC5A/RAI3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GPRC5A/7893
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias G protein-coupled receptor, family C, group 5, member A, GPCR5A, Orphan G-protein-coupling receptor PEIG-1, RAI3G-protein coupled receptor family C group 5 member A, RAIG1RAIG-1, retinoic acid induced 3, retinoic acid responsive, Retinoic acid-induced gene 1 protein, retinoic acid-induced protein 3
Host Species Mouse
Immunogen Recombinant fragment (around aa200-357) of human GPRC5A/RAI3 protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 9052
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.