missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPRC5B Recombinant Rabbit Monoclonal Antibody (8Y9J4)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555461
This item is not returnable.
View return policy
Description
Immunogen Sequence: TPNYFDTSQP RMRETAFEED VQLPRAYMEN KAFSMDEHNA ALRTAGFPNG SLGKRPSGSL GKRPSAPFRS NVYQPTEMAV VLNGGTIPTA PPSHTGRHLW.
This gene encodes a member of the type 3 G protein-coupled receptor family. Members of this superfamily are characterized by a signature 7-transmembrane domain motif. The encoded protein may modulate insulin secretion and increased protein expression is associated with type 2 diabetes. Alternative splicing results in multiple transcript variants.
Specifications
| GPRC5B | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q923Z0, Q9NZH0 | |
| GPRC5B | |
| A synthetic peptide corresponding to a sequence within amino acids 304-403 of human GPRC5B. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 8Y9J4 | |
| Unconjugated | |
| GPRC5B | |
| A-69G12.1; G protein-coupled receptor class C group 5 member B; G protein-coupled receptor, class C, group 5, member B; G protein-coupled receptor, family C, group 1, member B; GPRC5B; G-protein coupled receptor family C group 5 member B; RAIG2; RAIG-2; retinoic acid responsive gene protein; retinoic acid-induced gene 2 protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 293546, 51704, 64297 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction