missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPRC5C Antibody (577315) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6594Z
This item is not returnable.
View return policy
Description
GPRC5C Monoclonal antibody specifically detects GPRC5C in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| GPRC5C | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 577315 | |
| Flow Cytometry, CyTOF-ready | |
| G protein-coupled receptor, family C, group 5, member C, MGC131820, RAIG-3G-protein coupled receptor family C group 5 member C, RAIG3orphan G-protein coupled receptor, retinoic acid responsive gene protein, Retinoic acid-induced gene 3 protein | |
| NS0 mouse myeloma cell line transfected with recombinant human GPRC5C, Accession # Q9NQ84 | |
| 0.1 mL | |
| GPCR | |
| 55890 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction