missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPRIN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565173
This item is not returnable.
View return policy
Description
Immunogen sequence: EPEILGKGDPV APGRMDPMTV RKEDLGSLGK VDPLCSSKTY TVSPRKEDPG SLRKVDPVSS DKVDPVFPRK EEPRYSGKEH PVSSEKVAPT Highest antigen sequence identity to the following orthologs - mouse 45%, rat 29%.
May be involved in neurite outgrowth.
Specifications
| GPRIN1 | |
| Polyclonal | |
| Unconjugated | |
| GPRIN1 | |
| G protein regulated inducer of neurite outgrowth 1; G protein-regulated inducer of neurite outgrowth 1; Gprin1; GRIN1; KIAA1893; Z16 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114787 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z2K8 | |
| GPRIN1 | |
| Recombinant Human GPRIN1. Recombinant protein control fragment (Product #RP-104666). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction