missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPSM2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15790501 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15790501 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15790501 Supplier Invitrogen™ Supplier No. PA552555

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: FQSNRMDDQR CCLQEKNCHT ASTTTSSTPP KMMLKTSSVP VVSPNTDEFL DLLASSQSRR LDDQRASFSN LPGLRLTQNS QSVLSHLMTN DNKEADEDFF D Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 75%.

The protein encoded by this gene belongs to a family of proteins that modulate activation of G proteins, which transduce extracellular signals received by cell surface receptors into integrated cellular responses. The N-terminal half of this protein contains 10 copies of leu-gly-asn repeat, and the C-terminal half contains 4 GoLoco motifs, which are involved in guanine nucleotide exchange. This protein may play a role in neuroblast division and in the development of normal hearing. Mutations in this gene are associated with autosomal recessive nonsyndromic deafness.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPSM2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPSM2
Gene Accession No. P81274
Gene Alias 6230410J09Rik; CMCS; DFNB82; G protein signaling modulator 2; G-protein signaling modulator 2; G-protein signaling modulator 2 (AGS3-like, C. elegans); G-protein signalling modulator 2 (AGS3-like, C. elegans); G-protein-signaling modulator 2; GPSM2; HGNC:29501; hypothetical protein MGC63574; LGN; LGN protein; Mosaic protein LGN; Pins; Pins homolog; pins homolog (Drosophia); Pinsa; RGD1560967; TPR-motif and GoLoco-motif containing protein; zgc:63574
Gene Symbols GPSM2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPSM2. Recombinant protein control fragment (Product #RP-89660).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29899
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.