missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPT Polyclonal Antibody
GREENER_CHOICE

Product Code. 16380135
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16380135 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16380135 Supplier Invitrogen™ Supplier No. PA583179

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: FRGGYVEVVN MDAAVQQQML KLMSVRLCPP VPGQALLDLV VSPPAPTDPS FAQFQAEKQA VLAE Highest antigen sequence indentity to the following orthologs: Rat - 83%, Mouse - 81%.

GPT(Glutamate-pyruvate transaminase), also known as alanine aminotransferase, catalyzes the reversible conversion of L-alanine and alpha-ketoglutarate to L-glutamate and pyruvate. This protein has 2 distinct molecular and genetic forms: one cytoplasmic (soluble) (GPT1) and one mitochondrial (GPT2). See ALTQTL1 and ALTQTL2 for information on quantitative trait loci influencing the plasma level of alanine aminotransferase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPT
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPT
Gene Accession No. P24298
Gene Alias 1300007J06Rik; 2310022B03Rik; AAT1; alanine aminotransferase; alanine aminotransferase 1; ALT; ALT1; glutamate pyruvate transaminase 1; glutamic pyruvate transaminase; glutamic pyruvic transaminase (alanine aminotrasferase); glutamic pyruvic transaminase 1, soluble; glutamic pyruvic transaminase, soluble; glutamic-alanine transaminase 1; glutamic--alanine transaminase 1; glutamic-pyruvate transaminase (alanine aminotransferase); glutamic--pyruvic transaminase; glutamic--pyruvic transaminase 1; GPT; GPT 1; Gpt1; Gpt-1
Gene Symbols GPT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPT. Recombinant protein control fragment (Product #RP-93885).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2875
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.