missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPT2 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-26095UV
This item is not returnable.
View return policy
Description
GPT2 Polyclonal antibody specifically detects GPT2 in Rat samples. It is validated for ELISA
Specifications
| GPT2 | |
| Polyclonal | |
| ELISA | |
| AAT2, alanine aminotransferase 2, ALT2EC 2.6.1.2, Glutamate pyruvate transaminase 2, glutamic pyruvate transaminase (alanine aminotransferase) 2, Glutamic--alanine transaminase 2, glutamic-pyruvate transaminase 2, Glutamic--pyruvic transaminase 2, GPT 2 | |
| Rabbits immunized with purified, recombinant Rat GPT2 (Accession#: NP_001012057.1, Met1-Ser522) | |
| 0.1 mL | |
| metabolism | |
| 84706 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction