missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPVI Antibody (784808), Janelia Fluor™ 646, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB6758J
This item is not returnable.
View return policy
Description
GPVI Monoclonal antibody specifically detects GPVI in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| GPVI | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 51206 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 784808 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| Glycoprotein 6, glycoprotein VI (platelet), GPIV, GPVIplatelet collagen receptor, MGC138168, platelet glycoprotein VI | |
| Mouse myeloma cell line NS0-derived recombinant mouse GPVI, Gly24-Lys265, Accession # P0C191 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction