missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPX5 Monoclonal Antibody (5A1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA532925
This item is not returnable.
View return policy
Description
GPX5 Monoclonal Antibody for Western Blot, ICC/IF, Flow
Protects cells and enzymes from oxidative damage, by catalyzing the reduction of hydrogen peroxide, lipid peroxides and organic hydroperoxide, by glutathione. May constitute a glutathione peroxidase-like protective system against peroxide damage in sperm membrane lipids.
Specifications
| GPX5 | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.2% BSA, 50% Glycerol and 0.05% sodium azide; pH 7.4 | |
| O75715, P21765 | |
| GPX5 | |
| Synthetic peptide within Human GPX5 aa 85-134. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot, Immunocytochemistry, Western Blot | |
| 5A1 | |
| Unconjugated | |
| GPX5 | |
| Arep; arMEP24; AV379049; EGLP; epididymal androgen-related protein; epididymal secretory glutathione peroxidase; epididymis secretory sperm binding protein Li 75p; epididymis-specific glutathione peroxidase-like protein; Glutathione peroxidase 5; Gpx5; GPx-5; GSHPx-5; HEL-S-75p; lysosomal trafficking regulator, long splice form; major androgen-regulated protein; OTTHUMP00000017919; OTTHUMP00000214636 | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| 14780, 2880 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction