missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPX6 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

€ 208.00 - € 504.00

Specifications

Antigen GPX6
Concentration 0.5 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17750554
GREENER_CHOICE
View Documents
Invitrogen™
PA5119143
200 μL
€ 504.00
200µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17783344
View Documents
Invitrogen™
PA5121950
20 μL
€ 208.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GPX6 Polyclonal Antibody for Western Blot, IHC (P)

The protein encoded by this gene belongs to the glutathione peroxidase family, members of which catalyze the reduction of hydrogen peroxide, organic hydroperoxides and lipid hydroperoxides, and thereby protect cells against oxidative damage. Several isozymes of this gene family exist in vertebrates, which vary in cellular location and substrate specificity. Expression of this gene has been observed in embryos and olfactory epithelium; however, the exact function of this gene is not known. This isozyme is a selenoprotein in humans, containing the rare amino acid selenocysteine (Sec) at its active site. Sec is encoded by the UGA codon, which normally signals translation termination. The 3' UTRs of selenoprotein mRNAs contain a conserved stem-loop structure, designated the Sec insertion sequence (SECIS) element, that is necessary for the recognition of UGA as a Sec codon, rather than as a stop signal. The orthologs of this gene in mouse and rat (and some other species) contain a cysteine (Cys) residue in place of the Sec residue, and their corresponding mRNAs lack SECIS element.
Specifications

Specifications

GPX6
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Rat
Gpx6
Glutathione Peroxidase 6; Glutathione Peroxidase 6 Olfactory
Gpx6
Primary
Antigen affinity chromatography, Protein A
0.5 mg/mL
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q64625
259233
Recombinant protein Glutathione Peroxidase 6, Olfactory. The antigen corresponds to amino acid range 20-221 of the target protein.
Antibody
Videos
SDS
Documentos

Documentos

Certificados
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado