missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPX7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13501-1-AP-150UL
This item is not returnable.
View return policy
Description
Human GPX 7 is believed to be a member of the glutathione peroxidase family of proteins. These proteins play important roles in cellular defense mechanisms against oxidative damage. This is accomplished by catalyzing the reduction of hydroperoxides and hydrogen peroxide through the reducing substrate, glutathione.Specifications
| GPX7 | |
| Polyclonal | |
| Unconjugated | |
| GPX7 | |
| CL683, glutathione peroxidase 7, GPx 7, GPX6, GPX7, GSHPx 7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2882 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96SL4 | |
| GPX7 | |
| GPX7 Fusion Protein Ag4328 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction