Learn More
GPX7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13501-1-AP-20UL
 View more versions of this product
Product Code. 16841744
  • € 145.00 / 20µL
Description
Human GPX 7 is believed to be a member of the glutathione peroxidase family of proteins. These proteins play important roles in cellular defense mechanisms against oxidative damage. This is accomplished by catalyzing the reduction of hydroperoxides and hydrogen peroxide through the reducing substrate, glutathione.
Specifications
Specifications
GPX7
Polyclonal
Unconjugated
GPX7
CL683, glutathione peroxidase 7, GPx 7, GPX6, GPX7, GSHPx 7
Rabbit
Antigen Affinity Chromatography
RUO
2882
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96SL4
GPX7
GPX7 Fusion Protein Ag4328