Learn More
A&D Instruments GR Analytical balance Internal cal 310g x 0.1mg Class 1 Approved
GREENER_CHOICE
Brand:  A&D Instruments GR-300-EC
GR Analytical balance Internal cal 310g x 0.1mg Class 1 Approved
Product Code. 16641142
  • ONLINE EXCLUSIVE PRICE
    € 4320.75
    € 2819.82 / Each
    Save € 1500.93
    34% Off