missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GR/NR3C1 Antibody (BuGR2), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB300-731APC
This item is not returnable.
View return policy
Description
GR/NR3C1 Monoclonal specifically detects GR/NR3C1 in Human, Mouse, Rat, Guinea Pig, Rabbit, Sheep, Yeast samples. It is validated for Flow Cytometry.
Specifications
| GR/NR3C1 | |
| Monoclonal | |
| APC | |
| GCR, glucocorticoid receptor, GRGCCR, GRLNuclear receptor subfamily 3 group C member 1, nuclear receptor subfamily 3, group C, member 1, nuclear receptor subfamily 3, group C, member 1 (glucocorticoid receptor) | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Using enzymatic digestion analysis, has been shown to react with the undigested 97 kDa GR, a 17 kDa DNA-binding trypsin fragment, and a 45 kDa steroid- and DNA-binding chymotrypsin fragment. | |
| Store at 4C in the dark. | |
| IgG2a |
| Flow Cytometry | |
| BuGR2 | |
| Flow Cytometry | |
| NR3C1 | |
| Partially purified rat GR. | |
| 0.1 mL | |
| Phospho Specific, Signal Transduction | |
| 2908 | |
| Human, Mouse, Rat, Guinea Pig, Rabbit, Sheep, Yeast | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction