Learn More
Grace™ Alltech™ GracePure™ SPE Columns
Description
- Extensively tested and tightly controlled media
- Less than 2% bed weight variation
- Excellent value
Specifications
Specifications
| Base Material | Silica |
| Carbon Load | 17.1% |
| Column Capacity (Metric) | 6 mL |
| Column Phase | Reverse |
| Column Type | SPE |
| Bed Weight | 500 mg |
| For Use With (Application) | Cleanup, Concentration and Recovery of Chromatography Samples |
| Particle Size | 50 μm |
| Sorbent | C18-Max |
| Endcapped | Yes |
| Show More |