Learn More
Grace™ Alltech™ GracePure™ SPE Columns
Product Code. p-3750751
Click to view available options
Bed Weight:
100 mg
200 mg
500 mg
1000 mg
2000 mg
Column Capacity (Metric):
1 mL
3 mL
6 mL
20 mL
Sorbent:
Amino
C18-Low
C18-Max
Florisil
Unit Size:
Each
Product Code. 10639670
Brand: Grace™ 5138767
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Clean up and concentrate problematic samples
  • Extensively tested and tightly controlled media
  • Less than 2% bed weight variation
  • Excellent value
Specifications
Base Material Silica
Carbon Load 17.1%
Column Capacity (Metric) 6 mL
Column Phase Reverse
Column Type SPE
Bed Weight 500 mg
For Use With (Application) Cleanup, Concentration and Recovery of Chromatography Samples
Particle Size 50 μm
Sorbent C18-Max
Endcapped Yes
Pore Size 60 Å
Surface Area (Metric) 518 m2/g