Learn More
Grace™ GracePure™ SPE Florisil SPE Columns
Brand:  Grace™ 5141521
Additional Details : Weight : 0.30000kg
Product Code. 10764242
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
Magnesium Silicate
Normal
2000 mg
75 to 150 μm
60 Å
12 mL
SPE
Referenced in Many EPA Methods, Ideally Suited for Pesticides and Metals
Florisil