Learn More
Grace Davison™ GracePure™ Anion-X Ion-Exchange SPE Column
Product Code. 10318519
Click to view available options
Bed Weight:
500 mg
Unit Size:
Each
Product Code. 10318519
Brand: Grace Davison™ 5138755
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For subsequent HPLC or GC analysis
Solid phase extraction products for subsequent HPLC or GC analysis of environmental, biological, food and beverage samples
Specifications
Column Capacity (Metric) 3 mL
Bed Weight 500 mg
For Use With (Application) Subsequent HPLC or GC Analysis of Environmental, Biological, Food and Beverage Samples
Type GracePure Anion-X Ion-Exchange SPE Column