Learn More
Grace Davison™ GracePure™ C18-Low Reversed-Phase SPE Column
For subsequent HPLC or GC analysis
Brand:  Grace Davison™ 5141526
Additional Details : Weight : 0.30000kg
Product Code. 10680811
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Solid phase extraction products for subsequent HPLC or GC analysis of environmental, biological, food and beverage samples
Specifications
Specifications
20 mL
5000 mg
Subsequent HPLC or GC Analysis of Environmental, Biological, Food and Beverage Samples
GracePure C18-Low Reversed-Phase SPE Column
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers