Learn More
Grace Davison™ GracePure™ C18-Max Reversed-Phase SPE Column
Product Code. 10889140
Change view
Click to view available options
Bed Weight:
5000 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Weight unitSize
10889140 5000 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10889140 Supplier Grace Davison™ Supplier No. 5141524
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For subsequent HPLC or GC analysis
Solid phase extraction products for subsequent HPLC or GC analysis of environmental, biological, food and beverage samples
Specifications
Column Capacity (Metric) 20 mL
Bed Weight 5000 mg
For Use With (Application) Subsequent HPLC or GC Analysis of Environmental, Biological, Food and Beverage Samples
Type GracePure C18-Max Reversed-Phase SPE Column