Learn More
Grace Davison™ GracePure™ Diol Normal-Phase SPE Column
For subsequent HPLC or GC analysis
Brand: Grace Davison™ 5138773
Additional Details : Weight : 0.30000kg
Description
Solid phase extraction products for subsequent HPLC or GC analysis of environmental, biological, food and beverage samplesSpecifications
3 mL | |
500 mg | |
Subsequent HPLC or GC Analysis of Environmental, Biological, Food and Beverage Samples | |
GracePure Diol Normal-Phase SPE Column |