Learn More
Grace Davison™ GracePure™ Diol Normal-Phase SPE Column
For subsequent HPLC or GC analysis
Brand:  Grace Davison™ 5138773
Additional Details : Weight : 0.30000kg
Product Code. 10378129
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Solid phase extraction products for subsequent HPLC or GC analysis of environmental, biological, food and beverage samples
Specifications
Specifications
3 mL
500 mg
Subsequent HPLC or GC Analysis of Environmental, Biological, Food and Beverage Samples
GracePure Diol Normal-Phase SPE Column
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers