Learn More
Grace Davison™ GracePure™ Silica Normal Phase SPE Column
Description
Solid phase extraction products for subsequent HPLC or GC analysis of environmental, biological, food and beverage samples
Specifications
Specifications
| Column Capacity (Metric) | 20 mL |
| Bed Weight | 5000 mg |
| For Use With (Application) | Subsequent HPLC or GC Analysis of Environmental, Biological, Food and Beverage Samples |
| Type | GracePure Silica Normal-Phase SPE Column |