Learn More
Grace Davison™ GracePure™ Silica Normal Phase SPE Column
For subsequent HPLC or GC analysis
Brand:  Grace Davison™ 5138778
Additional Details : Weight : 0.30000kg
Product Code. 10530811
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Solid phase extraction products for subsequent HPLC or GC analysis of environmental, biological, food and beverage samples
Specifications
Specifications
3 mL
200 mg
Subsequent HPLC or GC Analysis of Environmental, Biological, Food and Beverage Samples
GracePure Silica Normal-Phase SPE Column