Learn More
Fisherbrand™ Grade 1106 Cellulose Technical Filter Paper
Used for viscous liquids and coarse particles.
Brand: Fisherbrand™
Additional Details : Weight : 0.01000kg
Description
- Thick smooth paper with large pore structure
Specifications
Folded Disc | |
50 to 80 μm | |
For syrups, animal fats, vegetable fats, margarines, Gelatin, dense oils, emulsions, essential oils | |
100 Pack | |
500 mm |
Cellulose | |
50 to 80 μm | |
1106 | |
0.47 mm | |
500 mm |