Learn More
Sartorius Stedim Biotech™ Grade 37/N Creped Paper for Qualitative and Technical Analyses, Discs
Sartorius Stedim Biotech™ Grade 37/N Creped Paper for Qualitative and Technical Analyses, Discs are suitable for analytical and technical purposes where fast flow rate is desirable.
Brand:  Sartorius Stedim Biotech™ FT-3-480-700
Additional Details : Weight : 5.19500kg
 View more versions of this product
Product Code. 15962950
  • € 498.00 / Pack of 100
Estimated Shipment: 12-06-2024
to see stock.
Add to basket
Description
Description
  • Made of refined pulp and cotton linters
  • Ash content 0.1 to 0.15%
  • Very fast filtering, wide pore
  • Wet-strengthened
Specifications
Specifications
135 g/m2
Cotton Linters, Refined Pulp
4 sec.
100/Pk.
700 mm
Disc
15 μm
37/N
0.5 mm