Learn More
Sartorius Stedim Biotech™ Grade 37/N Creped Paper for Qualitative and Technical Analyses, Folded
Sartorius Stedim Biotech™ Grade 37/N Creped Paper for Qualitative and Technical Analyses, Folded are suitable for analytical and technical purposes where fast flow rate is desirable.
Brand:  Sartorius Stedim Biotech™ FT-4-480-400
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15933010
  • € 374.00 / Pack of 100
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
Description
Description
  • Made of refined pulp and cotton linters
  • Ash content 0.1 to 0.15%
  • Very fast filtering, wide pore
  • Wet-strengthened
Specifications
Specifications
135 g/m2
Cotton Linters, Refined Pulp
4 sec.
0.5 mm
Folded
15 μm
37/N
400 mm