Learn More
Sartorius Stedim Biotech™ Grade 393 Quantitative Papers, Sheets
Sartorius Stedim Biotech™ Grade 393 Quantitative Papers, Sheets are suitable for quantitative and gravimetric analyses as well as for pressure or vacuum filtration.
Brand:  Sartorius Stedim Biotech™ FT-2-127-340440
Additional Details : Weight : 1.49600kg
 View more versions of this product
Product Code. 15922740
  • € 289.00 / Pack of 100
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
Description
Description
  • Made of 100% cotton linters with an alpha-cellulose content of < 98%
  • Acid-washed to make the papers ashless and achieve high purity (ash content ≤ 0.01 % according to DIN 540370)
  • Wet-strengthened
  • Color-coded box for easy selection
  • Very fine pore, very dense, very slow filtering
Specifications
Specifications
100 g/m2
100% Cotton Linters (<98% alpha-cellulose; ash-free)
300 sec.
100/Pk.
340 mm
Sheet
1 μm
393
0.17 mm
440 mm