Learn More
Sartorius™ Grade FT 55 Creped Surface Qualitative Filter Papers
Brand:  Sartorius™ FT-2-348-580580
Additional Details : Weight : 0.10000kg
Product Code. 10225851
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
Sheet
5μm/sec.
FT 55
0.15 mm
580 L x 580 Wmm
580 mm
Cotton Linters, Refined Pulp
Used for the rapid filtration of relatively coarse precipitates; because of their creped structure they provide a larger filtration area than smooth filter paper
100 Pack
Qualitative Filter Paper
580 mm