Learn More
Karl Hecht™ Erlenmeyer Flask, Standard Ground
Click to view available options
Capacity (Metric):
25 mL
50 mL
250 mL
500 mL
1000 mL
Closure Size:
NS 14/23
NS 29/32
NS 45/40
Unit Size:
Each
Description
- Duran™ borosilicate glass.
- Graduated.
- Standard ground.
- Reinforced ground neck marked with standard ground size.
- Without stopper.
Specifications
Specifications
| Capacity (Metric) | 25 mL |
| Material | Borosilicate Glass (3.3) |
| Shape | Erlenmeyer |
| Standard Taper Joint | NS 14/23 |
| Graduated | Yes |
| Closure Size | NS 14/23 |