Learn More
Witeg™ Graduated Erlenmeyer Flasks With Screw Cap
Product Code. p-8010345
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
3000 mL
5 L
Unit Size:
Each
Product Code. 10675771
Brand: Witeg™ 5 507 003
Only null left
Add to basket
Add to basket
Witeg™ Graduated Erlenmeyer Flasks With Screw Cap are graduated and designed for storage of culture media and mixing.
  • Borosilicate glass 3.3 construction
  • Graduated
  • Screw cap included
  • Available in a variety of volume capacities
Specifications
Capacity (Metric) 100 mL
Height (Metric) 105 mm
Includes Screw cap
Closure Type Screw Cap
Diameter (Metric) Outer 64 mm