Learn More
Bochem™ Graduated Measuring Jug
Click to view available options
Capacity (Metric):
500 mL
1000 mL
2000 mL
Descrizione
- Constructed of stianless steel
- Conical shape
- With base
Specifica
Base Style | Conical |
Diameter (Metric)Top | 115mm |
Capacity (Metric) | 500 mL |
Material | Stainless Steel (18/10) |
Height (Metric) | 115mm |
Includes | Base |
Increments | 100mL graduations |