missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herenz Heinz™ Graduated Pipette
Click to view available options
:
Color: White
Specifications
Specifications
| For Use With (Application) | ESR |
| Graduation Color | White |
| Length (Metric) | 300 mm |
| Type | Westergren Pipets |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction