missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRAF Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32229JF549
This item is not returnable.
View return policy
Description
GRAF Polyclonal antibody specifically detects GRAF in Human samples. It is validated for Western Blot
Specifications
| GRAF | |
| Polyclonal | |
| Western Blot | |
| FLJ42530, GRAFGTPase regulator associated with focal adhesion kinase pp125(FAK), KIAA0621rho GTPase-activating protein 26, Oligophrenin-1-like protein, OPHN1L1, OPHN1LGTPase regulator associated with focal adhesion kinase, Rho GTPase activating protein 26, Rho-type GTPase-activating protein 26 | |
| The immunogen this antibody was made to, maps to a region between residue 611 to 661 of human GTPase Regulator Associated with Focal Adhesion Kinase pp125(FAK) using the numbering given in entry NP_055886.1 (GeneID 23092). | |
| 0.1 mL | |
| Neuroscience, Stem Cell Markers | |
| 23092 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction