missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRAMD3 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97825AF594
This item is not returnable.
View return policy
Description
GRAMD3 Polyclonal specifically detects GRAMD3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GRAMD3 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| GRAM domain containing 3, GRAM domain-containing protein 3, HCV NS3-transactivated protein 2, hepatitis C virus nonstructural protein 3-transactivating protein 2, NS3TP2FLJ21313 | |
| Produced in rabbits immunized with E. coli-derived Human GRAMD3 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 65983 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction