missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRAMD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552673
This item is not returnable.
View return policy
Description
Immunogen sequence: GKRESKLGSA HSEAENGVEE KKKACRSPTA QSPTPSVEAD SPDQKKIISL WSKSSFDGAS LASDKNDCKT ESKNDPKTER KKSSSSSQYK ANMHFHKLFL SVPTEEPLKQ SFTCALQKEI LYQGKLFVSE NWI Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 83%.
Specifications
| GRAMD3 | |
| Polyclonal | |
| Unconjugated | |
| GRAMD2B | |
| 9030613F08Rik; 9130427A09Rik; AW061306; FLJ21313; GRAM domain containing 3; GRAM domain-containing protein 2B; GRAM domain-containing protein 3; GRAMD2B; GRAMD3; HCV NS3-transactivated protein 2; hepatitis C virus nonstructural protein 3-transactivating protein 2; NS3TP2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 65983 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96HH9 | |
| GRAMD2B | |
| Recombinant protein corresponding to Human GRAMD3. Recombinant protein control fragment (Product #RP-89852). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction