missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRAMD4 Antibody (PCRP-GRAMD4-1A10) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14282JF669
This item is not returnable.
View return policy
Description
GRAMD4 Monoclonal antibody specifically detects GRAMD4 in Human samples. It is validated for Flow Cytometry,Protein Array,Immunocytochemistry/ Immunofluorescence
Specifications
| GRAMD4 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| GRAM domain containing 4, GRAM domain-containing protein 4, KIAA0767 | |
| Recombinant full-length human GRAMD4 protein (Uniprot: Q6IC98) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Protein Array, Immunocytochemistry/Immunofluorescence | |
| PCRP-GRAMD4-1A10 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 23151 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction