Learn More
Grant Instruments™ Grant Bio™ Adapter Sets for R-12-10 Interchangeable Centrifuge Rotor
Product Code. p-8011786
Click to view available options
Capacity:
2 to 5 mL
4 to 8 mL
8 to 9 mL
Max. Tube Diameter:
13 mm
16 mm
Max. Tube Length:
75 mm
100 mm
Unit Size:
Each
Product Code. 15133706
Brand: Grant Instruments™ BN13100
Accommodates a variety of vacutainers for the R-12-10 Interchangabl;e Centrifuge Rotor
  • Interchangeable centrifuge rotor for 12 x 10 to 15mL tubes
  • Rounded ends, no caps
  • Dimensions: 16mm dia. x 105mm
TRUSTED_SUSTAINABILITY
Specifications
Capacity 4 to 8 mL
Max. Tube Diameter 13 mm
Max. Tube Length 100 mm
For Use With (Equipment) R-12-10
No. per Pack 12/Pk.