Learn More
Grant Instruments™ Grant Bio™ Deep Well Plates And Racks Tall Tubes Platform
Accommodates deepwell plates, microplates and tall tubes for Grant PTR-35 360° Vertical Multifunction Rotator.
Brand: Grant Instruments™ PRS-1DP
Additional Details : Weight : 0.50000kg
Description
- For use with PTR-35 360° Vertical Multifuntion Rotator
- Holds 1 deepwell plate, 1 microplate, or 1 rack for tall tubes 0.5 to 1.0mL
Specifications
Platform | |
For Microplates |