missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granulysin Antibody (B-R32), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18105DL594
This item is not returnable.
View return policy
Description
Granulysin Monoclonal antibody specifically detects Granulysin in Human samples. It is validated for ELISA
Specifications
| Granulysin | |
| Monoclonal | |
| DyLight 594 | |
| D2S69Elymphocyte-activation gene 2, granulysin, LAG2, LAG-2, Lymphokine LAG-2, NKG5519, Protein NKG5, TLA519T-cell activation protein 519, T-lymphocyte activation gene 519 | |
| Recombinant human Granulysin (NKG5 and LAG-2). Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 10578 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| ELISA | |
| B-R32 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction