missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granzyme A Antibody (CB9) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14619JF635
This item is not returnable.
View return policy
Description
Granzyme A Monoclonal antibody specifically detects Granzyme A in Human samples. It is validated for Flow Cytometry,Immunoprecipitation,Immunocytochemistry/ Immunofluorescence
Specifications
| Granzyme A | |
| Monoclonal | |
| Janelia Fluor 635 | |
| CTL tryptase, CTLA3Granzyme A (Cytotoxic T-lymphocyte-associated serine esterase-3; Hanukah factorserine protease), Cytotoxic T-lymphocyte proteinase 1, EC 3.4.21, fragmentin-1, granzyme A, granzyme A (granzyme 1, cytotoxic T-lymphocyte-associated serine esterase 3), Granzyme-1, H factor, Hanukkah factor, HF, HFSPEC 3.4.21.78 | |
| Full length human granzyme A | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Cancer, Immunology, Innate Immunity, Tumor Suppressors | |
| 3001 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunoprecipitation, Immunocytochemistry/Immunofluorescence | |
| CB9 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction