Learn More
Granzyme B Antibody (351927), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB2906CL1
Product Code. 30149748
  • € 1085.00 / 0.10mL
Description
Granzyme B Monoclonal antibody specifically detects Granzyme B in Human samples. It is validated for Immunohistochemistry,Western Blot,Simple Western,Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Granzyme B
Monoclonal
CoraFluor 1
C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTSGL1, granzyme B, GranzymeB, GZMB, HLP, SECT
Mouse myeloma cell line NS0-derived recombinant human Granzyme B, Gly19-Tyr247, Accession # P10144
0.1 mL
Apoptosis
3002
Store at 4°C in the dark. Do not freeze.
IgG2a
Immunohistochemistry, Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), Immunocytochemistry
351927