missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granzyme B Antibody (351927), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2906Y
This item is not returnable.
View return policy
Description
Granzyme B Monoclonal antibody specifically detects Granzyme B in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Specifications
| Granzyme B | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Simple Western, Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 351927 | |
| Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTSGL1, granzyme B, GranzymeB, GZMB, HLP, SECT | |
| Mouse myeloma cell line NS0-derived recombinant human Granzyme B, Gly19-Tyr247, Accession # P10144 | |
| 0.1 mL | |
| Apoptosis | |
| 3002 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction