Learn More
Granzyme B Antibody (351927), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB2906MFV610
Product Code. 18539827
  • € 674.00 / 0.10mL
Description
Granzyme B Monoclonal antibody specifically detects Granzyme B in Human samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Granzyme B
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
Mouse
0.1 mL
Apoptosis
3002
Store at 4C in the dark.
Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry
351927
Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTSGL1, granzyme B, GranzymeB, GZMB, HLP, SECT
Mouse myeloma cell line NS0-derived recombinant human Granzyme B, Gly19-Tyr247, Accession # P10144