Learn More
Granzyme B Antibody (CL10662) [Alexa Fluor™ 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-07984AF405
Product Code. 30294209
  • € 748.00 / 0.10mL
Description
Granzyme B Monoclonal antibody specifically detects Granzyme B in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Granzyme B
Monoclonal
Alexa Fluor 405
C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTSGL1, granzyme B, GranzymeB, GZMB, HLP, SECT
This antibody was developed using a recombinant protein derived from P10144, with the exact immunogen sequence remaining proprietary.
0.1 mL
Apoptosis
3002
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL10662
50mM Sodium Borate
Mouse
Protein A purified