Learn More
Granzyme B Antibody (GB11), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-64774PCP
Product Code. 18521132
  • ONLINE EXCLUSIVE PRICE
    € 754.00
    € 712.95 / 0.10mL
    Save € 41.05
    5% Off
Description
Granzyme B Monoclonal antibody specifically detects Granzyme B in Human, Monkey, Primate samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Granzyme B
Monoclonal
PerCP
PBS
Mouse
Protein A purified
RUO
Primary
Human, Monkey, Primate
Purified
Flow Cytometry
GB11
Flow Cytometry
C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTSGL1, granzyme B, GranzymeB, GZMB, HLP, SECT
Purified human Granzyme B (GZMB)
0.1 mL
Apoptosis
3002
Store at 4C in the dark.