missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granzyme B Antibody (OTI4E4), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70870R
This item is not returnable.
View return policy
Description
Granzyme B Monoclonal specifically detects Granzyme B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Granzyme B | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GZMB | |
| Recombinant protein fragment corresponding to amino acids 21-247 of human Granzyme B (NP_004122) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI4E4 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTSGL1, granzyme B, GranzymeB, GZMB, HLP, SECT | |
| Mouse | |
| Affinity Purified | |
| Apoptosis | |
| 3002.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction