missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granzyme D Antibody (RM0246-8G14), DyLight 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12409UV
This item is not returnable.
View return policy
Description
Granzyme D Monoclonal specifically detects Granzyme D in Mouse samples. It is validated for Western Blot.
Specifications
| Granzyme D | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Gzmd | |
| Mouse recombinant Granzyme D | |
| 0.1 mL | |
| 14941 | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot | |
| RM0246-8G14 | |
| Western Blot | |
| CCP5, Ctla5, Ctla-5, granzyme D | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction