missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granzyme G Antibody (RM0247-12f21), Allophycocyanin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12410APC
This item is not returnable.
View return policy
Description
Granzyme G Monoclonal antibody specifically detects Granzyme G in Mouse samples. It is validated for Western Blot
Specifications
| Granzyme G | |
| Monoclonal | |
| APC | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 14944 | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot | |
| RM0247-12f21 | |
| Western Blot | |
| Ctla7, Ctla-7 | |
| Mouse recombinant Granzyme G | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction